Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SMER28

Catalog No. 57312110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-312-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-312-110MG Supplier MilliporeSigma™ Supplier No. 57312110MG
Add to Cart
Add to Cart

A cell-permeable quinazoline compound that augments the cytostatic effects of Rapamycin and acts as an autophagy stimulator both in mammalian cultures

A cell-permeable quinazoline compound that augments the cytostatic effects of Rapamycin (Cat. Nos. 553210 and 553211) in Saccharomyces cerevisiae and acts as an autophagy stimulator both in mammalian cultures in vitro and in a Drosophila melanogaster HD model in vivo . Although SMER28 and Rapamycin do exhibit additive effects on the clearance of cellular autophagy substrates, SMER28 functions independently of Rapamycin by presumably acting on celluar target(s) downstream of the Rapamycin/FKBP12 target, mTOR.
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 95% by HPLC
CAS 307538-42-7
Molecular Formula C11H10BrN3
Formula Weight 264.1g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.