Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SNAPtide™ Botulinum Toxin A Substrate, Fluorogenic

Catalog No. 567333200NM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 nmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-733-3200NM 200 nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-733-3200NM Supplier MilliporeSigma™ Supplier No. 567333200NMOL
Add to Cart
Add to Cart

SNAPtide™ Botulinum Toxin A Substrate, Fluorogenic, is a synthetic peptide that acts as a FRET substrate with the fluorophore o-aminobenzoic acid and the acceptor chromophore 2,4-dinitrophenol (DNP).

A synthetic peptide, fluorescence resonance energy transfer (FRET) substrate containing the N-terminally-linked fluorophore o -aminobenzoic acid (Abz) and the acceptor chromophore 2,4-dinitrophenol (DNP) linked to the C-terminus. The substrate is based on the native botulinum toxin type A cleavage site found in the synaptosome-associated protein SNAP-25. Cleavage of the substrate by botulinum toxin releases the fluorophore and full fluorescence is restored.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from water.
Description 90% by HPLC
Target Proteins Substrate for the native botulinum toxin
Solubility DMSO
Form Lyophilized
Quantity 200 nmol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.