Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SNC80

Catalog No. 5081610001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-816-10001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-816-10001 Supplier MilliporeSigma™ Supplier No. 5081610001
Add to Cart
Add to Cart

A very potent and selective δ opioid receptor agonist (IC50

A very potent and selective δ opioid receptor agonist (IC50 = 0.32μM). Shown to induce dose- and time-dependent antinociception after i.c.v., i.th. and i.p. administration in mice. Widely used in studies of depression and inflammatory injuries. It is also reported to be acted upon heteromeric u-d receptors.
TRUSTED_SUSTAINABILITY

Specifications

Color Cream
Description 98% by HPLC
Target Proteins Dela opioid receptor
Molecular Formula C28H39N3O2
Solubility DMSO or 1N HCl
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.