Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Sodium Orthovanadate

Catalog No. 5675405GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-754-05GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-754-05GM Supplier MilliporeSigma™ Supplier No. 5675405GM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A broad spectrum potent inhibitor of protein tyrosine phosphatases.

A broad spectrum potent inhibitor of protein tyrosine phosphatases. Also known to inhibit Na+,K+-ATPase, acid and alkaline phosphatases, phosphofructokinase, and adenylate kinase. Also inhibits the ATPase activity of the reconstituted binding protein-dependent ATP-Binding Cassette (ABC) transporter for maltose (MalFGK2) of Salmonella typhimurium in the micromolar range. Can be converted to pervanadate by hydrogen peroxide.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins Na+,K+-ATPase, acid , alkaline phosphatases, phosphofructokinase, and adenylate kinase
CAS 13721-39-6
Molecular Formula Na3VO4
Formula Weight 183.9g/mol
Solubility H2O
Form Solid
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.