Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Soluble Epoxide Hydrolase Inhibitor, NCND

Catalog No. 32481325MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-481-325MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-481-325MG Supplier MilliporeSigma™ Supplier No. 32481325MG
Add to Cart
Add to Cart

The Soluble Epoxide Hydrolase Inhibitor, NCND, also referenced under CAS 402939-18-8, controls the biological activity of Soluble Epoxide Hydrolase.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 95% by TLC
Target Proteins Soluble epoxide hydrolase
Molecular Formula C19H38N2O
Formula Weight 310.5g/mol
Solubility CH2Cl2
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.