Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Somatostatin

Catalog No. 052308505MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-230-8505MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-230-8505MG Supplier MilliporeSigma™ Supplier No. 052308505MG

Cyclic tetradecapeptide.

Peptide Sequence: H-Ala-Gly-Cys3-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys14-OH (disulfide bond: 3 → 14)
TRUSTED_SUSTAINABILITY

Specifications

Description 98% by HPLC
Target Proteins Inhibits the release of growth hormone, insulin, and glucagon.
CAS 51110-01-1
Molecular Formula C76H104N18O19S2
Formula Weight 1637.9g/mol
Solubility 5% acetic acid
Content And Storage Hygroscopic
Form Lyophilized
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.