Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Spautin-1

Catalog No. 56756910MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-756-910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-756-910MG Supplier MilliporeSigma™ Supplier No. 56756910MG
Add to Cart
Add to Cart

A quinazolin compound that acts as a specific and potent inhibitor of autophagy and promotes Vps34 PI 3-kinase complex degradation by blocking the activity of USP 10 (IC50 = 580nM) and USP 13 (IC50 = 690nM) deubiquitinating enzymes

A quinazolin compound that acts as a specific and potent inhibitor of autophagy and promotes Vps34 PI 3-kinase complex degradation by blocking the activity of USP 10 (IC50 = 580nM) and USP 13 (IC50 = 690nM) deubiquitinating enzymes. However, it does not affect the lipid kinase activity of Vps34. Also shown to reduce the level of cytoplasmic and nuclear p53. Sensitizes breast cancer cell lines (BCAP37, MCF-7, and BT549) to apoptosis under conditions of nutritional deprivation. Also shown to inhibit etoposide-induced cell death in Bax/Bak double knockout (DKO) cells. Normal cell lines (e.g., MDCK cells) do not undergo autophagy in the presence of Spautin-1.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Molecular Formula C15H11F2N3
Formula Weight 271.26g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.