Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Sphingosine Kinase 1 Inhibitor II, PF-543

Catalog No. 5677415MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-774-15MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-774-15MG Supplier MilliporeSigma™ Supplier No. 5677415MG
Add to Cart
Add to Cart

The Sphingosine Kinase 1 Inhibitor II, PF-543 controls the biological activity of Sphingosine Kinase 1. This small molecule/inhibitor is primarily used for Cell Structure applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Molecular Formula C27H31NO4S
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.