Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Sphingosine Kinase 1 Inhibitor, SKI-178

Catalog No. 56773210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-773-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-773-210MG Supplier MilliporeSigma™ Supplier No. 56773210MG
Add to Cart
Add to Cart

A cell-permeable pyrazolocarbohydrazide compound that acts as a potent, reversible, sphingosine competitive and ATP-non-competitive inhibitor of sphingosine kinase 1 (SphK1) activity with excellent selectivity over SphK2 (Ki

A cell-permeable pyrazolocarbohydrazide compound that acts as a potent, reversible, sphingosine competitive and ATP-non-competitive inhibitor of sphingosine kinase 1 (SphK1) activity with excellent selectivity over SphK2 (Ki = 1.33 and >> 25μM, respectively). Shown to suppress sphingosine-1-phosphate (S1P) production in A549 cells and efficiently induce apoptosis in several tumor cells (IC50 in the range of 0.1-1.8μM).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 92% by HPLC
Molecular Formula C21H22N4O4
Formula Weight 394.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.