Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Sphingosine Kinase-2 Inhibitor, K145

Catalog No. 5091060001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-910-60001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-910-60001 Supplier MilliporeSigma™ Supplier No. 5091060001
Add to Cart
Add to Cart

A cell permeable, selective, sphingosine competitive inhibitor of spingosine Kinase -2 (IC50 = 4.3 μM).

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins Sphk2
Molecular Formula C18H24N2O3S
Solubility DMSO or H2O
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.