Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ sPLA2-IIA Inhibitor I

Catalog No. 5251451MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-514-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-514-51MG Supplier MilliporeSigma™ Supplier No. 5251451MG
Add to Cart
Add to Cart

The sPLA2-IIA Inhibitor I controls the biological activity of sPLA2-IIA. This small molecule/inhibitor is primarily used for Cell Signaling applications.

  • Packaged Under Inert Gas
  • Peptide Sequence: cyclic(2-NaphthylAla-Leu-Ser-2-NaphthylAla-Arg)•TFA
TRUSTED_SUSTAINABILITY

Specifications

Color White
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins sPLA2-IIA (human type IIA secreted phospholipase A2
Molecular Formula C41H50N8O6·CF3CO2H
Formula Weight 864.9g/mol
Solubility DMSO
Content And Storage Hygroscopic
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.