Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SPUTOLYSIN™ Reagent

Catalog No. 56000010ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-000-010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-000-010ML Supplier MilliporeSigma™ Supplier No. 56000010ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Intended for isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi, and yeasts from sputum

  • Concentrate of Dithiothreitol (DTT) in phosphate buffer, pH 6.5-7.5
  • Each vial is sufficient to make 100mL of working solution
TRUSTED_SUSTAINABILITY

Specifications

Formulation 20 vials each containing 10mL of concentrated DTT OR 1 vial containing 10mL of concentrated DTT
Molecular Weight (g/mol) 154.24
CAS 12-3-3483
Molecular Formula C4H10O2S2
Content And Storage 15°C to 30°C
Form Liquid
Quantity 10 mL
Product Type Calbiochem SPUTOLYSIN Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.