Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Src Inhibitor, PP1

Catalog No. 5678091MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-780-91MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-780-91MG Supplier MilliporeSigma™ Supplier No. 5678091MG
Add to Cart
Add to Cart

The Src Inhibitor PP1, also referenced under CAS 172889-26-8, controls the biological activity of Src. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

A cell-permeable pyrazolopyrimidine compound that is shown to inhibit Src family tyrosine kinases Lck, Fyn, Hck, and Src (IC50 = 5, 6, 20, and 170 nM, respectively), as well as non-Src family kinases such as CSK, RIP2, and CK1δ (IC50 = 640 nM, 26 nM, 170 nM, respectively), in in vitro kinase assays. At 1μM, in vitro kinase activity is reduced by more than 57% for p38α, p38β, CK1δ, CSK, RIP2, and GAK, while exhibiting much lower activity against more than 60 other kinases. Kinetic studies employing Src and [poly Glu-Tyr 4:1] indicate that the inhibition is mixed competitive with respect to the substrate and non-competitive with respect to ATP. PP1 also demonstrates anti-growth properties in several B lymphoma cultures of human and murine origins (IC50 = 2.5 to 8.0μM) and pediatric brain tumor cells (IC50 = 1.1 to 9.7μM). Also available in InSolution™ format (Cat. No. 539741).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 90% by HPLC
CAS 172889-26-8
Molecular Formula C16H19N5
Formula Weight 281.36g/mol
Solubility DMSO
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.