Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SRPK Inhibitor, SRPIN340

Catalog No. 5042930001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-429-30001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-429-30001 Supplier MilliporeSigma™ Supplier No. 5042930001
Add to Cart
Add to Cart

A cell-permeable isonicotinamide that acts as an ATP-competitive SRPK1-selective inhibitor (IC50

A cell-permeable isonicotinamide that acts as an ATP-competitive SRPK1-selective inhibitor (IC50 = 0.14 and 1.8μM, respectively, against mSRPK1 and mSRPK2) with much reduced activity against 143 other kinases. Shown to effectively counteract IGF-1-induced anti-angiogenic to pro-angiogenic VEGF isoforms switch both in cultures in vitro (1 h 10μM SRPIN340 prior to 12 h IGF-1 stimulation of PCIPs) and in a murine hypoxia-induced retinal neovascularization model in vivo (10 pmol/μL/eye intraocular SRPIN340 injection) by inhibiting PKC/SRPK signaling-dependent, alternate splicing factor ASF- (SF2; splicing factor 2) mediated VEGF pro-mRNA PSS (proximal splice site) selection. Suppresses RNA virus Sindbis propagation (IC50 = 60μM in Vero cultures) and HCV-JFH1 replication (1 & 10μM SRPIN340 in Huh7.5.1 cultures). Exhibits no toxicity toward rats (2 g/kg p.o. for 2 wks), CHO (5mg/mL for 24 h), or Huh7 (30μM for 48 h).
TRUSTED_SUSTAINABILITY

Specifications

Color Light Beige
Description 99% by HPLC
Molecular Formula C18H18F3N3O
Formula Weight 349.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.