Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ STAT3 Inhibitor V, Stattic

Catalog No. 57309925MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-309-925MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-309-925MG Supplier MilliporeSigma™ Supplier No. 57309925MG
Add to Cart
Add to Cart

STAT3 Inhibitor V, Stattic, CAS 19983-44-9, is a cell-permeable inhibitor of STAT3 cellular function that targets the STAT3-SH2 domain and prevents its association with upstream Kinases.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 95% by HPLC
Target Proteins STAT3
CAS 19983-44-9
Molecular Formula C8H5NO4S
Formula Weight 211.2g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.