Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ STAT5 Inhibitor II, IQDMA

Catalog No. 42029410MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-029-410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-029-410MG Supplier MilliporeSigma™ Supplier No. 42029410MG
Add to Cart
Add to Cart

The STAT5 Inhibitor II, IQDMA controls the biological activity of STAT5.

A cell-permeable indoloquinoline compound that exhibits anticancer activity and inhibits the growth of A549 lung cancer cells (IC50 = 8μM), as well as HL-60 (IC50 = 5μM) and K562 (IC50 = 8μM) leukemia cells. IQDMA-mediated apoptosis is shown to correlate with a downregulation of STAT5 pathway signaling in HL-60 and K562 cultures, while JNK and p38 signalings are shown to be activated in A549 cells upon IQDMA treatment.
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Description 95% by HPLC
Molecular Formula C19H20N4
Formula Weight 304.39g/mol
Solubility DMSO
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.