Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Stearoyl-CoA Desaturase 1 Inhibitor, MF-438

Catalog No. 56940610MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-940-610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-940-610MG Supplier MilliporeSigma™ Supplier No. 56940610MG
Add to Cart
Add to Cart

Stearoyl-CoA Desaturase 1 Inhibitor, MF-438, CAS 921605-87-0, is a cell-permeable inhibitor of Stearoyl-CoA Desaturase 1 (SCD1; IC50 of 2.3nM).

A cell-permeable, thiadiazole-pyridazine compound that acts as an inhibitor of Stearoyl-CoA Desaturase 1 (SCD1) with an IC50 of 2.3 nM. In vivo , it exhibits an ED50 of 1-3mg/kg, p.o., in mouse liver PD assays.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% by HPLC
Molecular Formula C19H18F3N5OS
Formula Weight 421.44g/mol
Solubility DMSO
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.