Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ SU11652

Catalog No. 572660500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-266-0500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-266-0500UG Supplier MilliporeSigma™ Supplier No. 572660500UG
Add to Cart
Add to Cart

A cell-permeable pyrrole-indolinone compound that acts as a potent, reversible, and ATP-competitive tyrosine kinase receptor and angiogenic inhibitor.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Description 95% by HPLC
Target Proteins PDGFRB
CAS 326914-10-7
Molecular Formula C22H27ClN4O2
Formula Weight 414.9g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.