Learn More
MilliporeSigma™ Calbiochem™ Nα-Tosyl-Lys Chloromethyl Ketone, Hydrochloride
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Color | Off-white |
| Product Type | Protease Inhibitor |
| Target Proteins | trypsin like proteases |
| CAS | 4238-41-9 |
| Molecular Formula | C14H21ClN2O3S·HCl |
| Formula Weight | 369.3g/mol |
| Solubility | Aqueous solutions (unstable above pH 6.0) |
| Content And Storage | Hygroscopic |
| Form | Solid |
| Quantity | 250 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.