Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ U0126

Catalog No. 6620051MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-200-51MG 1 mg
66-200-55MG 5 mg
66-200-510MG 10 mg
66-200-520MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 66-200-51MG Supplier MilliporeSigma™ Supplier No. 6620051MG
Add to Cart
Add to Cart

U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC50 = 72 nM) and MEK2 (IC50 = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins MEK1 and MEK2
CAS 109511-58-2
Molecular Formula C18H16N6S2·0.5C2H5OH
Formula Weight 403.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Purity 98% by HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.