Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Wiskostatin

Catalog No. 6815255MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-152-55MG 5 mg
68-152-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 68-152-55MG Supplier MilliporeSigma™ Supplier No. 6815255MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A cell-permeable N-alkylated carbazole derivative that selectively blocks actin filament assembly.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Inhibitor
Target Proteins N-WASP (neural Wiskott Aldrich syndrome protein)
CAS 253449-04-6
Molecular Formula C17H18Br2N2O
Formula Weight 426.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 95% by HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.