Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Trypsin Inhibitor, Soybean

Catalog No. 65035100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-035-100MG 100 mg
65-035-1GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 65-035-100MG Supplier MilliporeSigma™ Supplier No. 65035100MG
Add to Cart
Add to Cart

The Trypsin Inhibitor, Soybean, also referenced under CAS 9035-81-8, controls the biological activity of Trypsin. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

A reversible inhibitor of trypsin that binds to trypsin in a stoichiometric, pH-dependent manner (Kd = 500nM at pH 7.8).
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Target Proteins trypsin
CAS 9035-81-8
Formula Weight 20000g/mol
Solubility 67 mM potassium phosphate, pH 7.6
Content And Storage Hygroscopic, Protect from Moisture
Form Lyophilized
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.