Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ α2-Antiplasmin, Human Plasma

Catalog No. 178221100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-822-1100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-822-1100UG Supplier MilliporeSigma™ Supplier No. 178221100UG
Add to Cart
Add to Cart

A 452-amino acid glycoprotein present in plasma at about 70 μg/ml.

A 452-amino acid glycoprotein present in plasma at about 70μg/ml. Efficiently inhibits the plasminogen activator-induced lysis of fibrin clots. Forms a covalent complex with plasmin and inactivates it.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 200 mM NaCl, 20 mM Bis-Tris, pH 6.4.
Description 95% by SDS-PAGE
Target Proteins Plasminogen activator-induced lysis of fibrin clots
Formula Weight 70000g/mol
Solubility H2O
Form Solid
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.