Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Adenylyl Cyclase Type V Inhibitor, NKY80

Catalog No. 1168505MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-685-05MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-685-05MG Supplier MilliporeSigma™ Supplier No. 1168505MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Adenylyl Cyclase Type V Inhibitor, NKY80, also referenced under CAS 299442-43-6, controls the biological activity of Adenylyl Cyclase Type V.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Orange
Product Type Inhibitor
Target Proteins Adenylate cyclase (AC) type V isoform
CAS 299442-43-6
Molecular Formula C12H11N3O2
Formula Weight 229.2g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Purity 97% by HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.