Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Swinholide A, Theonella swinhoei

Catalog No. 57477610UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-477-610UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-477-610UG Supplier MilliporeSigma™ Supplier No. 57477610UG
Add to Cart
Add to Cart

A dimeric dilactone macrolide that is cytotoxic to a variety of cancer cell lines in vitro.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Description 98% by HPLC
Target Proteins Disrupts the actin cytoskeleton
Molecular Formula C78H132O20
Formula Weight 1389.9g/mol
Solubility EtOH or Acetone
Content And Storage Protect from light
Form Oil
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.