Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TACE Substrate IV, Fluorogenic

Catalog No. 6164071MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-640-71MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-640-71MG Supplier MilliporeSigma™ Supplier No. 6164071MG
Add to Cart
Add to Cart

An internally-quenched fluorogenic decapeptide substrate that is useful for continuous monitoring of TACE activity and high-throughput screening of TACE inhibitors (kcat =

An internally-quenched fluorogenic decapeptide substrate that is useful for continuous monitoring of TACE activity and high-throughput screening of TACE inhibitors (kcat = 21.6 s-1 and substrate concentration at half the Vmax, S0.5 = 342μM). The substrate is cleaved at Ala-Val amide bond. Acts as a poor substrate for MMP-1, MMP-9, and MMP-13. Displays better solubility in aqueous buffers.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Formulation Supplied as a trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Fluorogenic decapeptide substrate that is useful for continuous monitoring of TACE activity
Molecular Formula C59H93N23O20
Formula Weight 1444.5g/mol
Solubility DMSO
Content And Storage Protect from Light, Protect from Moisture
Form Lyophilized
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.