Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Tankyrase 1/2 Inhibitor VI, G007-LK

Catalog No. 5049070001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-490-70001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-490-70001 Supplier MilliporeSigma™ Supplier No. 5049070001
Add to Cart
Add to Cart

Tankyrase 1/2 Inhibitor VI, G007-LK, CAS 1380672-07-0, is a cell-permeable, potent and highly selective inhibitor of tankyrase (IC50 = 33 and 26nM for TNKS1 and TNKS2, respectively).

A cell-permeable, triazolyl-vinyl-oxadiazole compound that targets adenosine site of the NAD+ pocket in the TNKS PARP domain and acts as a potent and highly selective tankyrase inhibitor (IC50 = 33 and 26 nM, respectively, against TNKS1/PARP5a/ARTD5 and TNKS2/PARP5b/ARTD6 in auto-PARsylation assays), effectively inhibiting murine Wnt3a-induced reporter activity in human HEK293 and murine 10T1/2 cultures (ICmax ∽300 and 600 nM, respectively). Shown to display little or no inhibitory potency (IC50 >10μM) toward 7 other PARP enzymes (PARP1, 2, 3, 6, 7,10,11), 90 kinases, 16 phosphatases, and 75 GPCRs (IC50 >10μM). Shown to inhibit two CRC lines, COLO-320DM & SW403, colony formation in vitro (200nM) and tumor expansion in mice in vivo (20mg/kg via daily i.p.), although G007-LK toxicity is observed at higher dosages (≥30mg/kg/12 h or 60mg/kg/d via i.p.).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins Tankyrase 1/2
Molecular Formula C25H16ClN7O3S
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.