Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Tankyrase1/2 Inhibitor IV, JW55

Catalog No. 57554725MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-554-725MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-554-725MG Supplier MilliporeSigma™ Supplier No. 57554725MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Tankyrase1/2 Inhibitor IV, JW55 controls the biological activity of Tankyrase1/2. This small molecule/inhibitor is primarily used for Biochemicals applications.

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 99% HPLC
Target Proteins tankyrase 1 and 2
CAS 664993-53-7
Formula Weight 434.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.