Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Tankyrase1/2 Inhibitor V, WIKI4

Catalog No. 5005560001 Shop All MilliporeSigma Products
Change view
Click to view available options
Form:
Powder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
50-055-60001 Powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-055-60001 Supplier MilliporeSigma™ Supplier No. 5005560001
Add to Cart
Add to Cart

A cell-permeable triazole derivative that acts as a reversible TNKS inhibitor (IC50 = 15nM) and selectively inhibits Wnt/β-catenin, but not NF-κB, TGFβ, or retinoic acid, signaling pathway-dependent transcription activ

A cell-permeable triazole derivative that acts as a reversible TNKS inhibitor (IC50 = 15nM) and selectively inhibits Wnt/β-catenin, but not NF-κB, TGFβ, or retinoic acid, signaling pathway-dependent transcription activity. Both WIKI4 and XAV939 (Cat. No. 575545) are shown to induce basal axin1/2 protein accumulation in DLD1 colon cancer cells (2.5μM; 2 to 4 h) and prevent Wnt3a-induced axin1 protein degradation and β-catenin accumulation in A375 melanoma cells (1μM; up to 24 h) with similar potency. Likewise, WIKI4 also effectively inhibits the Wnt/β-catenin pathway-dependent growth of DLD1 (by 67% in 10 days;1μM).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
TRUSTED_SUSTAINABILITY

Specifications

Color Pale Yellow
Description 98% by HPLC
Target Proteins TNKS2
Molecular Formula C29H23N5O3S
Formula Weight 521.59g/mol
Solubility DMSO
Content And Storage Protect from light
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.