Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TAPI-2

Catalog No. 5790521MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-905-21MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-905-21MG Supplier MilliporeSigma™ Supplier No. 5790521MG
Add to Cart
Add to Cart

TAPI-2, CAS 187034-31-7, is a hydroxamate-based inhibitor of MMPs and TACE. Inhibits the activation-induced shedding of L-selectin from neutrophils, eosinophils, and lymphocytes.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins MMPs
CAS 187034-31-7
Molecular Formula C19H37N5O5
Formula Weight 415.5g/mol
Solubility EtOH or H2O
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.