Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Tau Aggregation Inhibitor, ATPZ, Cpd16

Catalog No. 58022210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-022-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 58-022-210MG Supplier MilliporeSigma™ Supplier No. 58022210MG
Add to Cart
Add to Cart

A water soluble (32uM in water) BBB-permeable ATPZ (aminothienopyridazine) tau aggregation inhibitor

A water soluble (32uM in water) BBB-permeable ATPZ (aminothienopyridazine) tau aggregation inhibitor that exhibited IC50 of 8.1uM in a heparin-induced tau assembly assay (monitored by thioflavine-T binding and fluorescence), and was further validated in vitro to reduce tau fibril formation (tau fragment K18PL, and full length tau40) by >50% in sedimentation assays. It also displayed very good B/P ratio (1.6) in a mice full PK study with good oral bioavailability and good metabolic stability. No cytotoxicity was observed at up to 100uM in HEK-293 cells.
TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Description 95% by HPLC
Molecular Formula C16H13ClN4O2S
Formula Weight 360.82g/mol
Solubility DMSO
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.