Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Tetrabenazine, TBZ

Catalog No. 5098920001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-989-20001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-989-20001 Supplier MilliporeSigma™ Supplier No. 5098920001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A monoamine-depleting agent that blocks vesicular monoamine transporter 2 activity. Promotes late-stage differentiation of Pdx1 + pancreatic progenitor cells into Ngn3 + endocrine precursors.

  • Packaged Under Inert Gas
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins VMAT2
CAS 58-46-8
Molecular Formula C19H27NO3
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.