Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TGF-β RI Kinase Inhibitor II

Catalog No. 6164522MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-645-22MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-645-22MG Supplier MilliporeSigma™ Supplier No. 6164522MG
Add to Cart
Add to Cart

TGF-β RI Kinase Inhibitor II, CAS 446859-33-2, is a cell-permeable, potent, reversible, ATP-competitive inhibitor of TGF-β R1 kinase (IC50 = 23 nM and 4 nM for ALK5 binding & auto-phosphorylation).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
Target Proteins ALKS binding
CAS 446859-33-2
Molecular Formula C17H13N5
Formula Weight 287.3g/mol
Solubility DMSO or MeOH
Content And Storage Protect from light
Form Solid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.