Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TGF-β RI Kinase Inhibitor V

Catalog No. 6164562MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-645-62MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-645-62MG Supplier MilliporeSigma™ Supplier No. 6164562MG

missing translation for 'validMainframeMsg'

The TGF-β RI Kinase Inhibitor V, also referenced under CAS 627536-09-8, controls the biological activity of TGF-β RI. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Light Yellow
Description 97% by HPLC
Target Proteins TGF-β RI/ALK5 Kinase
CAS 627536-09-8
Molecular Formula C17H10ClFN6
Formula Weight 352.8g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 2 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.