Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TGR5 Receptor Agonist, Cpd23g

Catalog No. 5058540001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-585-40001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-585-40001 Supplier MilliporeSigma™ Supplier No. 5058540001
Add to Cart
Add to Cart

A cell-permeable, orally available, phenoxypyrimidine carboxamide derivative that acts as a highly potent and selective agonist of Protein coupled receptor TGR5 (EC50 = 720 pM an

A cell-permeable, orally available, phenoxypyrimidine carboxamide derivative that acts as a highly potent and selective agonist of Protein coupled receptor TGR5 (EC50 = 720 pM and 6.2nM for human and mouse TGR5, respectively). Does not exhibit any significant affinity towards other related targets such as GPR40, GPR119, and GPR120. Shown to increase glucagon-like peptide-1 (GLP-1) secretion in a dose-dependent manner and significantly reduce blood glucose levels in db/db mice following a single oral dose (50mg/kg). Exhibits desirable pharmacokinetic properties in rodent models (t½ = 1.5 h, Cmax = 56 ng/ml; and AUC = 147 ng.h/ml following an oral dose of 5mg/kg).
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins TGR5
Molecular Formula C25H25N3O2
Solubility DMSO
Content And Storage Protect from light
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.