Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Thapsigargin

Catalog No. 5860051MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
58-600-51MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 58-600-51MG Supplier MilliporeSigma™ Supplier No. 5860051MG
Add to Cart
Add to Cart

Thapsigargin, CAS 67526-95-8, is a cell-permeable, tumor-promoting sesquiterpene lactone that releases calcium by non-competitvley inhibiting endoplasmic reticular Ca2+-ATPase (IC50 = 4-13 nM).

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Description 97% by TLC
Target Proteins endoplasmic reticular Ca2+ ATPase
CAS 67526-95-8
Molecular Formula C34H50O12
Formula Weight 650.8g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.