Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Thrombin Receptor Agonist

Catalog No. 605208500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-520-8500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-520-8500UG Supplier MilliporeSigma™ Supplier No. 605208500UG
Add to Cart
Add to Cart

The Thrombin Receptor Agonist, also referenced under CAS 137339-65-2, controls the biological activity of Thrombin Receptor. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Peptide Sequence: H-Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe-OH
TRUSTED_SUSTAINABILITY

Specifications

Color White to Off-white
Formulation Supplied as trifluoroacetate salt.
Description 95% by HPLC
Target Proteins Thrombin Receptor
CAS 137339-65-2
Formula Weight 1740g/mol
Solubility H2O or 5% Acetic Acid
Form Solid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.