Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Thrombin Substrate III, Fluorogenic

Catalog No. 60521125MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-521-125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-521-125MG Supplier MilliporeSigma™ Supplier No. 60521125MG
Add to Cart
Add to Cart

Sensitive fluorogenic substrate for the quantitative determination of thrombin.

Peptide Sequence: Benzoyl-Phe-Val-Arg-AMC, HCl
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a hydrochloride salt.
Description 98% by HPLC
Molecular Formula C37H43N7O6·HCl
Formula Weight 718.3g/mol
Solubility DMSO
Content And Storage Protect from moisture
Form Lyophilized
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.