Learn More
MilliporeSigma™ Calbiochem™ Thrombospondin, Human Platelets
Catalog No. 60522525UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
Catalog No. Quantity
60-522-525UG 25 μg
1 options
Catalog No. 60-522-525UG Supplier MilliporeSigma™ Supplier No. 60522525UG
Add to Cart
Add to Cart
Blocks FGF-induced neovascularization of rat cornea.
EMD Millipore Calbiochem™ Thrombospondin, Human Platelets is an angiogenesis inhibitor purified from human platelets.
  • Native thrombospondin
  • Inducer of platelet aggregation
  • Involved in regulation of cellular proliferation
  • Prepared from platelets that have been shown by certified tests to be negative for HBsAg for antibodies to HIV HCV
  • Lyophilized from 600mM NaCl, 20mM Tris, 1mM CaCl2, 20% sucrose, pH 7.8
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 μg
Content And Storage ≤-70°C