Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Tiagabine Hydrochloride

Catalog No. 5062360001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-623-60001 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-623-60001 Supplier MilliporeSigma™ Supplier No. 5062360001
Add to Cart
Add to Cart

A potent uptake inhibitor highly selective for GABA Transport-1 (IC50 = 67nM in vivo

A potent uptake inhibitor highly selective for GABA Transport-1 (IC50 = 67nM in vivo ). Clinically used as an anticonvulsant. Often used in studies related to anxiety disorders and neural damages resulted from epileptics.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Target Proteins GAT-1
Molecular Formula C20H25NO2S2·HCl
Solubility H2O
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.