Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TIMP-1, Human Neutrophil Granulocyte

Catalog No. 6120805UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-208-05UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-208-05UG Supplier MilliporeSigma™ Supplier No. 6120805UG
Add to Cart
Add to Cart

Native TIMP-1 from human granulocyte.

Native TIMP-1 from human granulocyte. Member of a family of inhibitors that participate in the activation and regulation of MMP activity. Forms a non-covalent stoichiometric complex with latent and active MMPs. Binds to pro-MMP-9 and MMP-9 via their C-terminal domains.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 200 mM NaCl, 50 mM Tris-HCl, 5 mM CaCl2, 1μM ZnCl2, 0.05% BRIJ™ 35 Detergent, 0.05% NaN3, pH 7.0.
Description 90% by SDS-PAGE
Target Proteins MMP activity
Formula Weight 28000g/mol
Content And Storage Avoid Freeze-Thaw Cycles
Form Liquid
Quantity 5 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.