Learn More
MilliporeSigma™ Calbiochem™ TIRAP Inhibitor Peptide, Cell-Permeable
Shop All MilliporeSigma ProductsDescription
- Packaged Under Inert Gas
- Peptide Sequence: H-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-Leu-Gln-Leu-Arg-Asp-Ala-Ala-Pro-Gly-Gly-Ala-Ile-Val-Ser-OH
- Cell Permeable
Specifications
Specifications
| Color | White |
| Formulation | Supplied as a trifluoroacetate salt. |
| Description | 97% by HPLC |
| Target Proteins | LPS-induced NF-κB activation, PKR phosphorylation, and JNK phosphorylation in RAW.kB cells |
| Molecular Formula | C163H268N52O38S |
| Formula Weight | 3596.3g/mol |
| Solubility | DMSO |
| Content And Storage | Hygroscopic, Protect from light |
| Form | Lyophilized |
| Quantity | 1 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.