Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TMB Solution

Catalog No. CL07100ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100mL
1000mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CL07100ML 100mL
CL071000ML 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CL07100ML Supplier MilliporeSigma™ Supplier No. CL07100ML
Add to Cart
Add to Cart

Validated for use in ELISA for detection of TMB Solution. MilliporeSigma™; Calbiochem™ TMB Solution is soluble, non-toxic, non-flammable horseradish peroxidase substrate.

  • Form: Liquid
  • Formulation: In a mildly acidic buffer with proprietary stabilizers, pH 3.55 ± 0.20
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
For Use With (Application) ELISA
Quantity 100mL
pH 3.55 ± 0.20
Product Type TMB Solution
Storage Requirements Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.