Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Topoisomerase IIα Inhibitor, TSC24

Catalog No. 61485210MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-485-210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-485-210MG Supplier MilliporeSigma™ Supplier No. 61485210MG
Add to Cart
Add to Cart

The Topoisomerase IIα Inhibitor, TSC24 controls the biological activity of Topoisomerase. This small molecule/inhibitor is primarily used for Cell Structure applications.

A cell-permeable, TSC (thiosemicarbazone) derivative that acts as a selective and ATP competitive Topoisomerase IIα inhibitor and Fe chelator. Independent of its Fe chelating ability, this compound is shown to block Topo II&alpha-catalyzed ATP hydrolysis by competing with ATP for binding to the ATPase domain of Topo IIα (Kd = 18μM). It also displays M phase cell-cycle arrest at 1μM in nocodazole-treated HT-29 cells. This compound demonstrates anti-proliferative activity, without distinct or obvious selectivity, towards HeLa, HL-60, SGC7901, and HT-29 cultures (IC50 = 90 nM, 50 nM, 30 nM, and 20 nM, respectively), and a broader panel of tumor cell lines with IC50 values in the nanomolar range.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
Molecular Formula C13H14N4S
Formula Weight 258.34g/mol
Solubility DMSO
Form Powder
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.