Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Transglutaminase 2 Inhibitor, ZDON

Catalog No. 6164675MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-646-75MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 61-646-75MG Supplier MilliporeSigma™ Supplier No. 6164675MG
Add to Cart
Add to Cart

A cell-permeable, peptide-based, irreversible inhibitor of transglutaminase 2 (TG2; IC50 = 150nM for recombinant TG2). Reacts with the active site cysteine of TG2.

A cell-permeable, peptide-based (Z- QVPL) irreversible inhibitor of transglutaminase 2 (TG2) (IC50 = 150nM for recombinant TG2) that acts by reacting with the active site cysteine of TG2. Exhibits far less potency against TG1 and TG3. Shown to significantly increase PGC-1a and cytochrome c mRNA levels in Q7 and Q111 cells (∽50 mM). Also shown to enhance cytochrome c promoter activity by 250% in Q111 cells. Exhibits protective effect in cells exposed to 3-nitropropionic acid and NMDA, however, it does not directly affect mitochondrial bioenergetics.
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 98% by HPLC
Molecular Formula C31H45N7O9
Solubility DMSO
Form Powder
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.