Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Trio N-Terminal RhoGEF Domain Inhibitor, ITX3

Catalog No. 64589010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-589-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 64-589-010MG Supplier MilliporeSigma™ Supplier No. 64589010MG
Add to Cart
Add to Cart

The Trio N-Terminal RhoGEF Domain Inhibitor, ITX3, also referenced under CAS 347323-96-0, controls the biological activity of Trio N-Terminal RhoGEF Domain. This small molecule/inhibitor is primarily used for Cell Structure applications.

A cell-permeable thiazolo-benzimidazolone compound that inhibits Trio N-terminal RhoGEF domain- (TrioN) mediated RhoG activation (IC50 = 76μM) and effectively reduces cellular Rac activation (by 80% with 50μM ITX3) caused by TrioN, but not GEF337 (RhoA GEF), Tiam1 (Rac GEF), or Vav2 (Rac1/RhoA/Cdc42 GEF), overexpression in HEK293T cells. Culture treatment of ITX at 100μM is shown to reduce NGF-induced neurite outgrowth of rat PC12 pheochromocytoma cells by 50% and block the formation of multinucleated myotubes by 78% in C2C12 mouse myoblasts cultures induced for differentiation, while the differentiation-associated Myogenin and Troponin T expressions in Trio knock-down C2C12 cells are not affected by ITX at the same concentration.
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
CAS 347323-96-0
Molecular Formula C22H17N3OS
Formula Weight 371.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.