Learn More
Description
EMD Millipore Calbiochem™ Troponin T, Human Heart binds tropomyosin, which regulates Ca2+-dependent interaction of actin myosin.
- Native troponin T
- Binds tropomyosin, which regulates the Ca2+-dependent interaction of actin myosin
- Prepared from tissue of individuals that have been shown by certified tests to be negative for HBsAg for antibodies to HIV HCV
- Lyophilized from 10mM HCl
- Assay: ≥95% by SDS-PAGE
Specifications
Specifications
| Quantity | 100 μg |
| Content And Storage | -20°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.