Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TRPC3 Channel Inhibitor III, Pyr10

Catalog No. 64849410MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-849-410MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 64-849-410MG Supplier MilliporeSigma™ Supplier No. 64849410MG
Add to Cart
Add to Cart

A cell-permeable pyrazole derivative that acts as a selective blocker of the transient receptor potential cation channel TRPC3-medated ROCE (receptor-operated Ca2+ entry) and inhibits Carbachol- (

A cell-permeable pyrazole derivative that acts as a selective blocker of the transient receptor potential cation channel TRPC3-medated ROCE (receptor-operated Ca2+ entry) and inhibits Carbachol- (Cat. No. 212385) induced Ca2+ entry in TRPC3-expressing HEK293 cells (IC50 = 0.72μM; by Fura-2), while exhibiting much reduced potency against Stim1 and Orai1-coupled CRAC (Ca2+ release-activated Ca2+) channel-mediated SOCE (store-operated Ca2+ entry) in RBL-2H3 cells (IC50 = 13.08μM; by Fura-2). A great complement to Pyr2 (Cat. No. 203890), Pyr 3 (Cat. No. 648490), and Pyr6 (Cat. No. 203891) in Ca2+ signaling studies.
TRUSTED_SUSTAINABILITY

Specifications

Color Orange-brown
Description 98% by HPLC
Target Proteins TRPC3
Molecular Formula C18H13F6N3O2S
Formula Weight 449.4g/mol
Solubility DMSO
Content And Storage Protect from light
Form Semi-solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.