Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TRPC3 Channel Inhibitor, Pyr3

Catalog No. 64849010MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-849-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 64-849-010MG Supplier MilliporeSigma™ Supplier No. 64849010MG
Add to Cart
Add to Cart

The TRPC3 Channel Inhibitor, Pyr3, also referenced under CAS 1160514-60-2, controls the biological activity of TRPC3 Channel. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Dark Brown
Description 95% by HPLC
CAS 1160514-60-2
Molecular Formula C16H11Cl3F3N3O3
Formula Weight 456.6g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.