Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ TRPM4 inhibitor, 9-Phenanthrol

Catalog No. 64849250MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-849-250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 64-849-250MG Supplier MilliporeSigma™ Supplier No. 64849250MG
Add to Cart
Add to Cart

The TRPM4 inhibitor, 9-Phenanthrol, also referenced under CAS 484-17-3, controls the biological activity of TRPM4. This small molecule/inhibitor is primarily used for Biochemicals applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Description 95% by HPLC
CAS 484-17-3
Molecular Formula C14H10O
Formula Weight 194.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.